accessibility menu, dialog, popup

UserName

CAS RN 866620-45-3

CH3NNHSH2NBr

CAS RN 866620-45-3

IUPAC Name: (3-bromo-5-methylpyridin-2-yl)thiourea
Synonyms: 3-bromo-5-methylpyridin-2-ylthiourea
Molecular Weight (g/mol): 246.13
Molecular Formula: C7H8BrN3S
InChi Key: YISGTNGIIXFOQT-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(NC(N)=S)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results