accessibility menu, dialog, popup

UserName

CAS RN 861208-16-4

IUPAC Name: ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Synonyms: ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 303.54
Molecular Formula: C10H8BrClN2O2
InChi Key: PEINUXVIHDZXNP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(Br)N2C=C(Cl)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results