accessibility menu, dialog, popup

UserName

CAS RN 86060-81-3

H3COHNS(R)NHOOOHO

CAS RN 86060-81-3

IUPAC Name: (2R)-3-[(acetamidomethyl)sulfanyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2R)-3-[(acetamidomethyl)sulfanyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 414.48
Molecular Formula: C21H22N2O5S
InChi Key: CSMYOORPUGPKAP-IBGZPJMESA-N
SMILES: CC(=O)NCSC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results