CAS RN 86-59-9
CAS RN 86-59-9
IUPAC Name:
quinoline-8-carboxylic acid
Synonyms:
8-quinolinecarboxylic acid
Molecular Weight (g/mol):
173.17
Molecular Formula:
C10H7NO2
InChi Key:
QRDZFPUVLYEQTA-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C2N=CC=CC2=CC=C1