accessibility menu, dialog, popup

UserName

CAS RN 85118-02-1

H2NOFF

CAS RN 85118-02-1

IUPAC Name: 2,4-difluorobenzamide
Synonyms: 2,4-difluorobenzamide
Molecular Weight (g/mol): 157.12
Molecular Formula: C7H5F2NO
InChi Key: KTXFXDMDYZIXSJ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results