CAS RN 851086-95-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ent NAP 226-90, TRC
CAS: 851086-95-8 Molecular Formula: C10 H15 N O Molecular Weight (g/mol): 165.23 Synonym: Phenol, 3-[(1R)-1-(dimethylamino)ethyl]-,3-[(1R)-1-(Dimethylamino)ethyl]phenol,(R)-3-[1-(Dimethylamino)ethyl]phenol IUPAC Name: 3-[(1R)-1-(dimethylamino)ethyl]phenol SMILES: C[C@@H](N(C)C)c1cccc(O)c1