accessibility menu, dialog, popup

UserName

CAS RN 85068-36-6

IUPAC Name: (2,5-difluorophenyl)(phenyl)methanone
Synonyms: (2,5-difluorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 218.2
Molecular Formula: C13H8F2O
InChi Key: HSCUAAMDKDZZKG-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(=C1)C(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results