accessibility menu, dialog, popup

UserName

CAS RN 850589-53-6

BFOCH3HOOH

CAS RN 850589-53-6

IUPAC Name: (3-ethoxy-5-fluorophenyl)boronic acid
Synonyms: 3-ethoxy-5-fluorophenylboronic acid
Molecular Weight (g/mol): 183.97
Molecular Formula: C8H10BFO3
InChi Key: OVVBLFYHZAMJIK-UHFFFAOYSA-N
SMILES: CCOC1=CC(F)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results