accessibility menu, dialog, popup

UserName

CAS RN 850589-34-3

BONHOCH3OHHO

CAS RN 850589-34-3

IUPAC Name: {4-[(2-methoxyethyl)carbamoyl]phenyl}boronic acid
Synonyms: 4-[(2-methoxyethyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 223.04
Molecular Formula: C10H14BNO4
InChi Key: QRGVHOKYRIQQTN-UHFFFAOYSA-N
SMILES: COCCNC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results