accessibility menu, dialog, popup

UserName

CAS RN 850568-68-2

BNOHOOH

CAS RN 850568-68-2

IUPAC Name: [4-(5,6-dihydro-4H-1,3-oxazin-2-yl)phenyl]boronic acid
Synonyms: 4-(5,6-dihydro-4H-1,3-oxazin-2-yl)phenylboronic acid
Molecular Weight (g/mol): 205.02
Molecular Formula: C10H12BNO3
InChi Key: OASSAUSIPZASKK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=NCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results