accessibility menu, dialog, popup

UserName

CAS RN 850568-09-1

BOCH3H3CCH3HOOH

CAS RN 850568-09-1

IUPAC Name: [3-methyl-4-(propan-2-yloxy)phenyl]boronic acid
Synonyms: 4-isopropoxy-3-methylphenylboronic acid
Molecular Weight (g/mol): 194.04
Molecular Formula: C10H15BO3
InChi Key: XRGBZDZYYQVOJZ-UHFFFAOYSA-N
SMILES: CC(C)OC1=C(C)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results