accessibility menu, dialog, popup

UserName

CAS RN 845885-88-3

CH3NSNOOH3CCH3H3COHO

CAS RN 845885-88-3

IUPAC Name: 2-(2-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}-5-methyl-1,3-thiazol-4-yl)acetic acid
Synonyms: {2-[1-(tert-butoxycarbonyl)piperidin-4-yl]-5-methyl-1,3-thiazol-4-yl}acetic acid
Molecular Weight (g/mol): 340.44
Molecular Formula: C16H24N2O4S
InChi Key: IHHZQULPYDMAOP-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)N=C(S1)C1CCN(CC1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)