accessibility menu, dialog, popup

UserName

H3COHNNHO

CAS RN 83803-96-7

IUPAC Name: N'-(3-methylbenzoyl)naphthalene-2-carbohydrazide
Synonyms: N'-(3-methylbenzoyl)naphthalene-2-carbohydrazide
Molecular Weight (g/mol): 304.35
Molecular Formula: C19H16N2O2
InChi Key: NOZDWMYGSUBAEE-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)NNC(=O)C1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results