accessibility menu, dialog, popup

UserName

CAS RN 83265-56-9

IUPAC Name: 2-amino-5-(trifluoromethoxy)benzoic acid
Synonyms: 2-amino-5-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 221.14
Molecular Formula: C8H6F3NO3
InChi Key: UXNGDCBPIGOZFO-UHFFFAOYSA-N
SMILES: NC1=C(C=C(OC(F)(F)F)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results