accessibility menu, dialog, popup

UserName

CAS RN 796061-08-0

BOOH3CCH3H3CH3CNHSOOCH3

CAS RN 796061-08-0

IUPAC Name: 4-methyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzene-1-sulfonamide
Synonyms: 4-methyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide
Molecular Weight (g/mol): 373.27
Molecular Formula: C19H24BNO4S
InChi Key: RGAZOXRZFCGICF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results