CAS RN 791587-67-2
CAS RN 791587-67-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
c-Fms-IN-2, MedChemExpress
MedChemExpress c-Fms-IN-2 is a FMS kinase inhibitor with an IC50 of 0.024 μM.