accessibility menu, dialog, popup

UserName

CAS RN 791-29-7

H3CSi

CAS RN 791-29-7

IUPAC Name: methyltriphenylsilane
Synonyms: methyltriphenylsilane
Molecular Weight (g/mol): 274.44
Molecular Formula: C19H18Si
InChi Key: GIGVICQLYWGMGW-UHFFFAOYSA-N
SMILES: C[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results