accessibility menu, dialog, popup

UserName

CAS RN 7790-46-7

IIIIPt

CAS RN 7790-46-7

IUPAC Name: platinum(4+) tetraiodide
Synonyms: platinum(4+) tetraiodide
Molecular Weight (g/mol): 702.7
Molecular Formula: I4Pt
InChi Key: RNJPWBVOCUGBGY-UHFFFAOYSA-J
SMILES: [I-].[I-].[I-].[I-].[Pt+4]
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results