accessibility menu, dialog, popup

UserName

CAS RN 7783-39-3

FFHg

CAS RN 7783-39-3

IUPAC Name: mercury(2+) difluoride
Synonyms: mercury(II) fluoride mercury(I) fluoride
Molecular Weight (g/mol): 238.59
Molecular Formula: F2Hg
InChi Key: FMSYTQMJOCCCQS-UHFFFAOYSA-L
SMILES: [F-].[F-].[Hg++]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results