accessibility menu, dialog, popup

UserName

NaNaOSeOO

CAS RN 7782-82-3

IUPAC Name: disodium selenite
Synonyms: sodium selenite disodium selenite natriumselenit
Molecular Weight (g/mol): 172.95
Molecular Formula: Na2O3Se
InChi Key: BVTBRVFYZUCAKH-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-][Se]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)