accessibility menu, dialog, popup

UserName

CAS RN 77716-11-1

CH3H3CCH3OOHNNOHOCH3

CAS RN 77716-11-1

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}-1-methyl-1H-pyrrole-2-carboxylic acid
Synonyms: 4-[(tert-butoxycarbonyl)amino]-1-methylpyrrole-2-carboxylic acid
Molecular Weight (g/mol): 240.26
Molecular Formula: C11H16N2O4
InChi Key: GOLAEMFBWAIGRX-UHFFFAOYSA-N
SMILES: CN1C=C(NC(=O)OC(C)(C)C)C=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)