accessibility menu, dialog, popup

UserName

CAS RN 77326-62-6

BrNH2N

CAS RN 77326-62-6

IUPAC Name: 2-amino-6-bromobenzonitrile
Synonyms: 2-amino-6-bromobenzonitrile
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: ZKZYTSGUONPOPF-UHFFFAOYSA-N
SMILES: NC1=C(C#N)C(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results