accessibility menu, dialog, popup

UserName

CAS RN 773131-96-7

NNH2OOH(E/Z)

CAS RN 773131-96-7

IUPAC Name: 3-(2-aminopyridin-3-yl)prop-2-enoic acid
Synonyms: 3-(2-aminopyridin-3-yl)prop-2-enoic acid
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: HKUVFKTUMTYXQT-UHFFFAOYSA-N
SMILES: NC1=C(C=CC(O)=O)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results