accessibility menu, dialog, popup

UserName

CAS RN 7727-18-6

ClVClClO

CAS RN 7727-18-6

IUPAC Name: vanadium oxytrichloride
Synonyms: vanadium oxytrichloride
Molecular Weight (g/mol): 176.31
Molecular Formula: Cl3H3OV
InChi Key: JNKMSLLEOBTKQY-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.O=[V]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results