accessibility menu, dialog, popup

UserName

CAS RN 772-33-8

NOOBrOH

CAS RN 772-33-8

IUPAC Name: 2-(bromomethyl)-4-nitrophenol
Synonyms: 2-(bromomethyl)-4-nitrophenol
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: KFDPCYZHENQOBV-UHFFFAOYSA-N
SMILES: OC1=C(CBr)C=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results