accessibility menu, dialog, popup

UserName

HONNNFF

CAS RN 76674-21-0

IUPAC Name: 1-(2-fluorophenyl)-1-(4-fluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethan-1-ol
Synonyms: impact sopra flutriafol (PP450)
Molecular Weight (g/mol): 301.3
Molecular Formula: C16H13F2N3O
InChi Key: JWUCHKBSVLQQCO-UHFFFAOYNA-N
SMILES: OC(CN1C=NC=N1)(C1=CC=C(F)C=C1)C1=CC=CC=C1F
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results