accessibility menu, dialog, popup

UserName

CAS RN 762-84-5

CH3ONHCH3CH3H3C

CAS RN 762-84-5

IUPAC Name: N-tert-butylacetamide
Synonyms: N-tert-butylacetamide
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: ACYFWRHALJTSCF-UHFFFAOYSA-N
SMILES: CC(=O)NC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results