CAS RN 762-62-9
CAS RN 762-62-9
IUPAC Name:
4,4-dimethylpent-1-ene
Synonyms:
1-pentene, 4,4-dimethyl-
Molecular Weight (g/mol):
98.19
Molecular Formula:
C7H14
InChi Key:
KLCNJIQZXOQYTE-UHFFFAOYSA-N
SMILES:
CC(C)(C)CC=C
No Refinements
1
–
0
of
0
results
1
4,4-Dimethyl-1-pentene, 99%
CAS: 762-62-9 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00026337 InChI Key: KLCNJIQZXOQYTE-UHFFFAOYSA-N Synonym: 4,4-dimethyl-1-pentene,1-pentene, 4,4-dimethyl,2,2-dimethyl-4-pentene,4,4-dimethyl-pent-1-ene,4,4-dimethyl pent-1-ene,neoheptaene,acmc-1bcwv,ch3 3cch2ch=ch2,4,4'-dimethyl-1-pentene PubChem CID: 12984 IUPAC Name: 4,4-dimethylpent-1-ene SMILES: CC(C)(C)CC=C