CAS RN 754226-39-6
CAS RN 754226-39-6
IUPAC Name:
2-(3,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(3,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
240.06
Molecular Formula:
C12H15BF2O2
InChi Key:
VJTCPZZOSKBSCP-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC(F)=C(F)C=C1
1
–
1
of
1
results
1
3,4-Difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 754226-39-6 Molecular Formula: C12H15BF2O2 Molecular Weight (g/mol): 240.057 InChI Key: VJTCPZZOSKBSCP-UHFFFAOYSA-N Synonym: 2-3,4-difluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,1,3,2-dioxaborolane, 2-3,4-difluorophenyl-4,4,5,5-tetramethyl,acmc-209oye,3,4-difluorophenylboronic acid pinacol ester,3,4-difluorobenzeneboronic acid pinacol ester,2-3,4-difluoro-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane PubChem CID: 53429043 IUPAC Name: 2-(3,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)F