CAS RN 75139-39-8
CAS RN 75139-39-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carbazomycin A, TRC
CAS: 75139-39-8 Molecular Formula: C16H17NO2 Molecular Weight (g/mol): 255.31 Synonym: 3,4-Dimethoxy-1,2-dimethyl-9H-carbazole IUPAC Name: 3,4-dimethoxy-1,2-dimethyl-9H-carbazole SMILES: CC1=C(C)C(NC2=C3C=CC=C2)=C3C(OC)=C1OC