accessibility menu, dialog, popup

UserName

CAS RN 75-81-0

ClClBrBr

CAS RN 75-81-0

IUPAC Name: 1,2-dibromo-1,1-dichloroethane
Synonyms: 1,2-dibromo-1,1-dichloroethane
Molecular Weight (g/mol): 256.75
Molecular Formula: C2H2Br2Cl2
InChi Key: FIYBYNHDEOSJPL-UHFFFAOYSA-N
SMILES: ClC(Cl)(Br)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)