accessibility menu, dialog, popup

UserName

CH3CH3CH3CCH3CH3CH3CH3CH3CCH3CH3Mg

CAS RN 74507-64-5

IUPAC Name: magnesium(2+) bis(pentamethylcyclopenta-2,4-dien-1-ide)
Synonyms: magnesium(2+) bis(pentamethylcyclopenta-2,4-dien-1-ide)
Molecular Weight (g/mol): 294.77
Molecular Formula: C20H30Mg
InChi Key: QZNMZKZCVOJICG-UHFFFAOYSA-N
SMILES: [Mg++].C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results