CAS RN 73177-67-0
IUPAC Name:
2-methyl-1-phenylpentan-1-ol
Synonyms:
2-methyl-1-phenylpentan-1-ol
Molecular Weight (g/mol):
178.28
Molecular Formula:
C12H18O
InChi Key:
AKMRXUUGAJOMFO-UHFFFAOYNA-N
SMILES:
CCCC(C)C(O)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Methyl-1-phenyl-1-pentanol, erythro + threo, 97%, Thermo Scientific™
CAS: 73177-67-0 Molecular Formula: C12H18O Molecular Weight (g/mol): 178.275 MDL Number: MFCD00049211 InChI Key: AKMRXUUGAJOMFO-UHFFFAOYSA-N Synonym: 2-methyl-1-phenyl-1-pentanol,2-methyl-1-phenyl-1-pentanol, erythro + threo,2-pentyl phenyl carbinol,2-methyl-1-phenyl-1-pentanol, threo + erythro PubChem CID: 144583 IUPAC Name: 2-methyl-1-phenylpentan-1-ol SMILES: CCCC(C)C(C1=CC=CC=C1)O