CAS RN 728-86-9
CAS RN 728-86-9
IUPAC Name:
phenyl[4-(trifluoromethyl)phenyl]methanone
Synonyms:
phenyl[4-(trifluoromethyl)phenyl]methanone
Molecular Weight (g/mol):
250.22
Molecular Formula:
C14H9F3O
InChi Key:
OHTYZZYAMUVKQS-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
4-(Trifluoromethyl)benzophenone, 98%, Thermo Scientific™
CAS: 728-86-9 Molecular Formula: C14H9F3O Molecular Weight (g/mol): 250.22 MDL Number: MFCD00000400 InChI Key: OHTYZZYAMUVKQS-UHFFFAOYSA-N Synonym: 4-trifluoromethyl benzophenone,4-trifluoromethylbenzophenone,phenyl 4-trifluoromethyl phenyl methanone,p-trifluoromethylbenzophenone,phenyl-4-trifluoromethyl phenyl methanone,methanone, phenyl 4-trifluoromethyl phenyl,fxffr dvr,phenyl 4-trifluoromethyl phenyl ketone,acmc-20amto PubChem CID: 69767 IUPAC Name: phenyl-[4-(trifluoromethyl)phenyl]methanone SMILES: FC(F)(F)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1