accessibility menu, dialog, popup

UserName

CAS RN 72200-76-1

NH2HOOHHONH2HOOHHOOHOOHO(Z)

CAS RN 72200-76-1

IUPAC Name: (2Z)-but-2-enedioic acid; bis(2-amino-2-(hydroxymethyl)propane-1,3-diol)
Synonyms: maleic acid; bis(tris buffer)
Molecular Weight (g/mol): 358.34
Molecular Formula: C12H26N2O10
InChi Key: POSZUTFLHGNLHX-KSBRXOFISA-N
SMILES: NC(CO)(CO)CO.NC(CO)(CO)CO.OC(=O)\C=C/C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results