accessibility menu, dialog, popup

UserName

CAS RN 722-56-5

ONOOI

CAS RN 722-56-5

IUPAC Name: diphenyliodanium nitrate
Synonyms: diphenyliodanium nitrate
Molecular Weight (g/mol): 343.12
Molecular Formula: C12H10INO3
InChi Key: CQZCVYWWRJDZBO-UHFFFAOYSA-N
SMILES: [O-][N+]([O-])=O.[I+](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results