accessibility menu, dialog, popup

UserName

CAS RN 72093-04-0

CH3NCl

CAS RN 72093-04-0

IUPAC Name: 3-chloro-4-methylpyridine
Synonyms: 3-chloro-4-methylpyridine
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: VLNXNJMHLZFECV-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results