accessibility menu, dialog, popup

UserName

NHNSOOOOSOOONOONaNa(E/Z)

CAS RN 7143-21-7

IUPAC Name: disodium 4-[2-(4-nitrophenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium 4-[2-(4-nitrophenyl)hydrazin-1-ylidene]-3-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 497.36
Molecular Formula: C16H9N3Na2O9S2
InChi Key: QMNPQXAVQXMXTD-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-][N+](=O)C1=CC=C(NN=C2C(=O)C(=CC3=C2C=CC(=C3)S([O-])(=O)=O)S([O-])(=O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results