accessibility menu, dialog, popup

UserName

CAS RN 71255-09-9

H3CONO

CAS RN 71255-09-9

IUPAC Name: 2-methoxypyridine-3-carbaldehyde
Synonyms: 2-methoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: PIFFMIDNNWOQLK-UHFFFAOYSA-N
SMILES: COC1=NC=CC=C1C=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)