accessibility menu, dialog, popup

UserName

CAS RN 7116-96-3

IUPAC Name: 4-pentyl-1,1'-biphenyl
Synonyms: 4-pentyl-1,1'-biphenyl
Molecular Weight (g/mol): 224.35
Molecular Formula: C17H20
InChi Key: IFUOTAQBVGAZPR-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results