accessibility menu, dialog, popup

UserName

CAS RN 7045-70-7

CH3H3CCH3H3C

CAS RN 7045-70-7

IUPAC Name: 1,3-bis(propan-2-yl)cyclohexane
Synonyms: 1,3-diisopropylcyclohexane
Molecular Weight (g/mol): 168.32
Molecular Formula: C12H24
InChi Key: WDTCMYUFBNCSKK-UHFFFAOYNA-N
SMILES: CC(C)C1CCCC(C1)C(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results