accessibility menu, dialog, popup

UserName

CAS RN 70421-66-8

IUPAC Name: 2-[(E)-[2-(4-nitrophenyl)hydrazin-1-ylidene]methyl]pyridine
Synonyms: 2-[(E)-[2-(4-nitrophenyl)hydrazin-1-ylidene]methyl]pyridine
Molecular Weight (g/mol): 242.24
Molecular Formula: C12H10N4O2
InChi Key: JXFHYUCJPGWCFI-NTEUORMPSA-N
SMILES: [O-][N+](=O)C1=CC=C(N\N=C\C2=CC=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results