accessibility menu, dialog, popup

UserName

CAS RN 702640-48-0

CH3ClNH2F

CAS RN 702640-48-0

IUPAC Name: 6-chloro-2-fluoro-3-methylaniline
Synonyms: 6-chloro-2-fluoro-3-methylaniline
Molecular Weight (g/mol): 159.59
Molecular Formula: C7H7ClFN
InChi Key: QXHIYPYDZBWKFU-UHFFFAOYSA-N
SMILES: CC1=C(F)C(N)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results