accessibility menu, dialog, popup

UserName

CAS RN 6972-71-0

CH3NH2H3CNOO

CAS RN 6972-71-0

IUPAC Name: 4,5-dimethyl-2-nitroaniline
Synonyms: 4,5-dimethyl-2-nitroaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: PINGKGKKUSYUAW-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(C=C1C)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)