accessibility menu, dialog, popup

UserName

CAS RN 6950-92-1

CH3CH3OHH3C

CAS RN 6950-92-1

IUPAC Name: 2-(2,4,6-trimethylphenyl)ethan-1-ol
Synonyms: 2-(2,4,6-trimethylphenyl)ethanol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: FQZPTDPHVFTOSY-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CCO)C(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results