accessibility menu, dialog, popup

UserName

NH2ClFFF

CAS RN 69411-05-8

IUPAC Name: 3-chloro-5-(trifluoromethyl)aniline
Synonyms: 3-chloro-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 195.57
Molecular Formula: C7H5ClF3N
InChi Key: OVENUGPMQDFGLE-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results