CAS RN 69227-51-6
IUPAC Name:
1-ethyl-1-methylpyrrolidin-1-ium bromide
Synonyms:
1-ethyl-1-methylpyrrolidin-1-ium bromide
Molecular Weight (g/mol):
194.12
Molecular Formula:
C7H16BrN
InChi Key:
KHJQQUGSPDBDRM-UHFFFAOYSA-M
SMILES:
[Br-].CC[N+]1(C)CCCC1
1
–
1
of
1
results
1
1-Ethyl-1-methylpyrrolidinium bromide, 98%
CAS: 69227-51-6 Molecular Formula: C7H16BrN Molecular Weight (g/mol): 194.116 MDL Number: MFCD03095384 InChI Key: KHJQQUGSPDBDRM-UHFFFAOYSA-M Synonym: 1-ethyl-1-methylpyrrolidinium bromide,1-ethyl-1-methylpyrrolidin-1-ium bromide,1-ethyl-1-methylpyrrolidiniumbromide,pyrrolidinium, 1-ethyl-1-methyl-, bromide,1-methyl-1-ethylpyrrolidinium bromide,pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1,dsstox_cid_29306,dsstox_rid_83422,dsstox_gsid_49347,ksc495s4d PubChem CID: 112264 IUPAC Name: 1-ethyl-1-methylpyrrolidin-1-ium;bromide SMILES: CC[N+]1(CCCC1)C.[Br-]