accessibility menu, dialog, popup

UserName

CAS RN 690631-96-0

BOOH3CCH3H3CH3CNSFFFCH3

CAS RN 690631-96-0

IUPAC Name: 4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
Synonyms: 4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
Molecular Weight (g/mol): 369.21
Molecular Formula: C17H19BF3NO2S
InChi Key: ZUROXICAUJQTIB-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=C(C=C1)C(F)(F)F)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results