CAS RN 6835-13-8
CAS RN 6835-13-8
IUPAC Name:
2-{4-[(4-chloro-2-methylphenyl)carbamoyl]phenoxy}propanoate
Synonyms:
2-{4-[(4-chloro-2-methylphenyl)carbamoyl]phenoxy}propanoate
Molecular Weight (g/mol):
332.76
Molecular Formula:
C17H15ClNO4
InChi Key:
HEFAXWHJDHPLEJ-UHFFFAOYNA-M
SMILES:
CC(OC1=CC=C(C=C1)C(=O)NC1=CC=C(Cl)C=C1C)C([O-])=O
1
–
1
of
1
results
1
N-(n-Octyl)-n-pentylamine, 98+%, Thermo Scientific™
CAS: 6835-13-8 Molecular Formula: C17H15ClNO4 Molecular Weight (g/mol): 332.76 MDL Number: MFCD03093075 InChI Key: HEFAXWHJDHPLEJ-UHFFFAOYNA-M Synonym: n-n-octyl-n-pentylamine,pentyloctylamine,octyl pentyl amine,n-n-octyl-n-pentylamine, 98+% PubChem CID: 20069586 SMILES: CC(OC1=CC=C(C=C1)C(=O)NC1=CC=C(Cl)C=C1C)C([O-])=O