accessibility menu, dialog, popup

UserName

CAS RN 67911-21-1

H3COOOH3C

CAS RN 67911-21-1

IUPAC Name: 6,6-dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
Synonyms: 6,6-dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: QKAHKEDLPBJLFD-UHFFFAOYNA-N
SMILES: CC1(C)C2C1C(=O)OC2=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results